Assay Details
| Gene Symbol: RDH13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096798.1 | - | 1 | 355 | 153 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000586845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000586845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000586845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000586845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000586961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000586961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000586961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012045.hg.1 | ENST00000586331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012045.hg.1 | ENST00000586331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012045.hg.1 | ENST00000586331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012045.hg.1 | ENST00000586331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012045.hg.1 | ENST00000586331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012045.hg.1 | ENST00000586331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012045.hg.1 | ENST00000586331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012045.hg.1 | ENST00000586331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012045.hg.1 | ENST00000586331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012045.hg.1 | ENST00000586331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012045.hg.1 | ENST00000592423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000586845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000586845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000586845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000586845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000586961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000586961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000586961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008886.hg.1 | ENST00000593060.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 123965 | - | - |

RDH13
Genomic Map