Assay Details
| Gene Symbol: CCL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032965.5 | NP_116741.2 | 4 | 122 | ||
| GenBank mRNA | AI244164.1 | - | 1 | 95 | 122 | |
| U58914.1 | - | 4 | 121 | |||
More Information
Updated Mapping Information
Set Membership: |
|

CCL15
Genomic Map