Assay Details
| Gene Symbol: LOC646522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120444.1 | 3 | 674 | 75 | ||
| GenBank mRNA | BC043577.2 | - | 2 | 647 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | opioid binding protein/cell adhesion molecule like | OPCML |
IGLON1, OBCAM, OPCM |
-251.2kb | Sense | Chr. 11 - 132403361 - 133532983 on NCBI Build GRCh38 |
| Downstream | spermatogenesis associated 19 | SPATA19 |
CT132, SPAS1, spergen1 |
51.4kb | Sense | Chr. 11 - 133835930 - 133845538 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138575 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138587 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566372 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566236 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138564 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138648 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566352 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138583 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566237 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138570 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566238 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16519678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138672 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566324 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138657 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138558 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138537 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566235 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566250 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566305 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138580 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566275 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566284 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566234 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566356 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138692 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138547 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138634 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138568 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566337 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138577 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566340 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566375 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138533 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566369 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566382 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138639 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138574 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566343 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138593 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138637 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138652 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566318 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566392 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138526 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566246 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566270 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138598 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138655 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566361 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566264 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566304 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138621 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138589 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138553 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138695 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138551 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16519684 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566242 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566341 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16519690 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138569 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566338 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566396 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138687 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16519679 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138530 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566260 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138654 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566332 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138697 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566273 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566335 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138651 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566308 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566233 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566268 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138636 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566309 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138616 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138538 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138550 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566319 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138675 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138664 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16519680 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138591 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138638 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138606 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566321 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138602 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566241 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566269 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138670 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138599 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566288 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138679 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138620 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566299 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138677 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566251 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138576 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566368 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566389 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138548 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566322 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566394 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566255 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566248 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566297 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566376 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138611 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138646 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566360 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138566 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138635 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138628 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566307 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566393 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566306 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138640 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566240 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138565 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566254 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566258 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138542 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138680 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08138624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28566325 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 78920 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 144924 | AK097332.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 144924 | BC032536.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 627811 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 498261 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 869273 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 350500 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 114607 | AK122900.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 114607 | AK172762.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 114607 | AK307755.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 114607 | AY092062.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 114607 | AY134855.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 114607 | BC111059.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 114607 | BC117279.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 114607 | NM_152994.2 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 475828 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 522194 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 919871 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 261480 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 153245 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 243910 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3301489 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 39166 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02553343 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P19427448 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P02553343 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P02553343 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P19427477 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P19427448 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02553343 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 97503 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 147843 | AF493888.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 147843 | AK096447.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 147843 | AK223502.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 147843 | BC002530.2 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 147843 | BT006674.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 147843 | L13687.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 147843 | NM_001667.2 | refseq_mrna_human_NM |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 90148 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02553329 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19427477 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19427448 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19427458 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19427490 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02553343 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P19099565 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19427492 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19427448 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19427477 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02553343 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1563013_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 237662_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 237662_at | - | - |
| Human Genome U133 Plus 2.0 Array | 237662_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1563013_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1563013_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563013_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237662_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237662_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237662_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563013_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563013_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237662_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563013_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237662_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563013_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237662_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237662_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563013_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563013_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563013_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563013_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237662_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237662_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237662_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563013_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237662_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563013_at5 | - | - |
| Human Genome U95 Set | 68724_at | - | - |
| Human Genome U95 Set | 68724_PM_at | - | - |
| Human Genome U133 Set | 237662_PM_at | - | - |
| Human Genome U133 Set | 237662_at | - | - |

LOC646522
Genomic Map