Assay Details
| Gene Symbol: PIK3CD-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027045.1 | 2 | 859 | 60 | ||
| GenBank mRNA | AK097325.1 | - | 2 | 868 | 60 | |
| BC016063.1 | - | 2 | 838 | 60 | ||
| BC144613.1 | - | 1 | 987 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002184.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3789056 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4957 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4956 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15801 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3789056 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3789056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038065 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20048363 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20048354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038063 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20048370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038042 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038054 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15021949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038050 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038054 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038063 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15021949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038050 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038065 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20048354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20048370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038042 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20048363 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00038048 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P709232 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038053 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038068 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038045 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038037 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038042 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038029 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15021940 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038056 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038052 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038019 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038031 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038061 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15021949 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038025 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038047 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038043 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038071 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15021942 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038059 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038050 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038023 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038073 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038032 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038054 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038033 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038021 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038038 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15021938 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038069 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038036 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038027 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00038065 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7912256 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7912255 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7912254 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1559638_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012811.hg.1 | ENST00000377320.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012811.hg.1 | ENST00000377320.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012811.hg.1 | NR_027045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012811.hg.1 | NR_027045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033627.hg.1 | ENST00000377320.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033627.hg.1 | NR_027045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033627.hg.1 | TC01002184.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033628.hg.1 | ENST00000377320.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033628.hg.1 | NR_027045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033628.hg.1 | TC01002184.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100194679.hg.1 | ENST00000377320.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194679.hg.1 | NR_027045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194679.hg.1 | ENST00000377320.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194679.hg.1 | NR_027045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194679.hg.1 | TC0100012811.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100194678.hg.1 | ENST00000377320.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194678.hg.1 | NR_027045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194678.hg.1 | ENST00000377320.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194678.hg.1 | NR_027045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194678.hg.1 | TC0100012811.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012811.hg.1 | ENST00000377320.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012811.hg.1 | ENST00000377320.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012811.hg.1 | NR_027045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012811.hg.1 | NR_027045.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1559638_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1559638_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559638_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559638_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559638_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559638_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559638_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559638_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559638_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559638_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559638_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559638_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1559638_at7 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053940 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053940 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 232204 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1703119 | BC016063.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1703119 | BC144613.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1703119 | NR_027045.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1703119 | AK097325.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2157091 | BC016063.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2157091 | BC144613.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2157091 | NR_027045.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2157091 | AK097325.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1703119 | BC016063.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1703119 | BC144613.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1703119 | NR_027045.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1703119 | AK097325.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1703119 | BC016063.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1703119 | BC144613.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1703119 | NR_027045.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1703119 | AK097325.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2157091 | BC016063.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2157091 | BC144613.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2157091 | NR_027045.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2157091 | AK097325.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1703119 | BC016063.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1703119 | BC144613.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1703119 | NR_027045.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1703119 | AK097325.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1703119 | BC016063.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1703119 | BC144613.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1703119 | NR_027045.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1703119 | AK097325.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2157091 | BC016063.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2157091 | BC144613.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2157091 | NR_027045.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2157091 | AK097325.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2157091 | BC016063.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2157091 | BC144613.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2157091 | NR_027045.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2157091 | AK097325.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1703119 | BC016063.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1703119 | BC144613.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1703119 | NR_027045.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1703119 | AK097325.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1703119 | BC016063.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1703119 | BC144613.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1703119 | NR_027045.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1703119 | AK097325.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2157091 | BC016063.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2157091 | BC144613.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2157091 | NR_027045.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2157091 | AK097325.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1703119 | BC016063.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1703119 | BC144613.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1703119 | NR_027045.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1703119 | AK097325.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2157091 | BC016063.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2157091 | BC144613.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2157091 | NR_027045.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2157091 | AK097325.1 | human_transcriptome |

PIK3CD-AS1
Genomic Map