Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 7848 | MIR7848 | hsa-mir-7848 | -21.0kb | Anti-sense | Chr. 8 - 133046481 - 133046581 on NCBI Build GRCh38 |
| Downstream | WNT1 inducible signaling pathway protein 1 | WISP1 |
CCN4, WISP1c, WISP1i, WISP1tc |
121.0kb | Sense | Chr. 8 - 133191039 - 133231690 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map