| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010365.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010365.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010365.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010365.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010365.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010365.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010365.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010365.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010365.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010365.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000412934.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000457681.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000460965.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011232.hg.1 |
ENST00000463223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010365.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010365.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010365.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010365.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010365.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010365.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010365.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010365.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010365.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010365.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300175550.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300175550.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300175550.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300175550.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300175550.hg.1 |
TC0300010365.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300175552.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300175552.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300175552.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300175552.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300175552.hg.1 |
TC0300010365.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300175553.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300175553.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300175553.hg.1 |
ENST00000502417.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300175553.hg.1 |
NR_110135.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300175553.hg.1 |
TC0300010365.hg.1 |
Clariom D TC |