Assay Details
| Gene Symbol: LOC101926983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135571.1 | 2 | 547 | 132 | ||
| GenBank mRNA | BC032918.2 | - | 2 | 550 | 132 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101926983
Genomic Map