Assay Details
| Gene Symbol: MAN1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098129.1 | - | 1 | 588 | 128 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1525 | LINC01525 | lnc-MAN1A2-1 | -205.2kb | Sense | Chr. 1 - 117295466 - 117321336 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100996263 | LOC100996263 | 68.0kb | Anti-sense | Chr. 1 - 117596832 - 117605770 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |

MAN1A2
Genomic Map