Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 19 member A2, C-C motif chemokine like |
FAM19A2 |
TAFA-2, TAFA2 |
Anti-sense | Chr. 12 - 61708248 - 62192839 on NCBI Build GRCh38 | |
| Downstream | ubiquitin specific peptidase 15 | USP15 |
UNPH-2, UNPH4 |
26.7kb | Sense | Chr. 12 - 62260340 - 62409721 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map