Assay Details
| Gene Symbol: ZC3H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011526669.2 | XP_011524971.1 | 1 | 110 | 139 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011526669.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000253048.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000594019.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000595341.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000597069.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | ENST00000601973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011014.hg.1 | NM_015168.1 | Refseq |

ZC3H4
Genomic Map