Assay Details
| Gene Symbol: ABCB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163942.1 | NP_001157414.1 | 6 | 1160 | 115 | |
| GenBank mRNA | BX452500.1 | - | 1 | 152 | 115 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ABCB5
Genomic Map