Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022411.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022411.hg.1 |
ENST00000540252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022411.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022411.hg.1 |
TC19001646.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022410.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022410.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022410.hg.1 |
TC19001646.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010984.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010984.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010984.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010984.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022408.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022408.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022408.hg.1 |
TC19001646.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900010984.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010984.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010984.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010984.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010984.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191425.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191425.hg.1 |
NM_032040.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900191425.hg.1 |
TC1900010984.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900191425.hg.1 |
ENST00000307522.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900191425.hg.1 |
NM_032040.4 |
Refseq |