Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transcription factor 25 | TCF25 |
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049 |
-3.0kb | Sense | Chr. 16 - 89873583 - 89911384 on NCBI Build GRCh38 |
| Downstream | melanocortin 1 receptor | MC1R |
CMM5, MSH-R, SHEP2 |
1.4kb | Sense | Chr. 16 - 89917879 - 89920977 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map