Assay Details
| Gene Symbol: TMEM63C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020431.3 | NP_065164.2 | 17 - 18 | 1742 | 103 | |
| GenBank mRNA | AK124749.1 | - | 3 - 4 | 250 | 103 | |
| AK125159.1 | - | 17 - 18 | 1674 | 103 | ||
| BC136613.1 | - | 17 - 18 | 1624 | 9020603 | 103 | |
| BC136614.1 | - | 17 - 18 | 1621 | 9020604 | 103 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14002202.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006199.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006199.hg.1 | ENST00000536110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006199.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006199.hg.1 | TC14002202.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006200.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006200.hg.1 | ENST00000536110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006200.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006200.hg.1 | TC14002202.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554346.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000554766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000555588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000556514.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | ENST00000557504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010635.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180234.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180234.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180234.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180234.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180234.hg.1 | TC1400010635.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400180233.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180233.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180233.hg.1 | ENST00000298351.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180233.hg.1 | NM_020431.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180233.hg.1 | TC1400010635.hg.1 | Clariom D TC |

TMEM63C
Genomic Map