Assay Details
| Gene Symbol: PMCHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003922.1 | 1 - 2 | 230 | 150 | ||
| GenBank mRNA | AY008412.1 | - | 1 - 2 | 230 | 150 | |
| AY008413.1 | - | 1 - 2 | 230 | 150 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724392 | LOC102724392 | Anti-sense | Chr. 5 - 71321031 - 71446756 on NCBI Build GRCh38 | ||
| Downstream |
B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB |
BDP1 |
HSA238520, TAF3B1, TFC5, TFIIIB'', TFIIIB150, TFIIIB90, TFNR |
77.8kb | Sense | Chr. 5 - 71455615 - 71578288 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000331.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05004445.hg.1 | ENST00000450213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05004445.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05004445.hg.1 | TC05000331.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000457791.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000457791.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000523548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000523548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | ENST00000523548.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000423102.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000423102.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000430902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000430902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000430902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000430902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | ENST00000430902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | NR_003921.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | NR_003921.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | NR_003921.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006935.hg.1 | NR_003921.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05004444.hg.1 | ENST00000450213.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05004444.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05004444.hg.1 | TC05000331.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500152501.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152501.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152501.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152501.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152501.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152501.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152501.hg.1 | TC0500007722.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500152500.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152500.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152500.hg.1 | ENST00000457791.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152500.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152500.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152500.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152500.hg.1 | ENST00000457791.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152500.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152500.hg.1 | TC0500007722.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000418902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000423102.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000423102.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000423301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000430902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000430902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000430902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000430902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | ENST00000430902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | NR_003921.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | NR_003921.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | NR_003921.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006935.hg.1 | NR_003921.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152498.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152498.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152498.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152498.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152498.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152498.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152498.hg.1 | TC0500007722.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500152499.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152499.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152499.hg.1 | ENST00000457791.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152499.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152499.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152499.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152499.hg.1 | ENST00000457791.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500152499.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500152499.hg.1 | TC0500007722.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000415808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000450213.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000457791.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000457791.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000523548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000523548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | ENST00000523548.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | NR_003922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007722.hg.1 | NR_003922.1 | Refseq |

PMCHL2
Genomic Map