Assay Details
| Gene Symbol: GPR179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004334.3 | NP_001004334.3 | 9 - 10 | 1918 | 123 | |
| GenBank mRNA | AY584611.1 | - | 9 - 10 | 1918 | 123 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001424.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17018844.hg.1 | ENST00000342292.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17018844.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17018844.hg.1 | TC17001424.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000610867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000610867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000610867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000610867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000622573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000622573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | ENST00000622573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700182780.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182780.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182780.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700182780.hg.1 | TC1700010518.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700182780.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182780.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182780.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000610867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000610867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000610867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000610867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000616987.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000621958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000622573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000622573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | ENST00000622573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010518.hg.1 | NM_001004334.3 | Refseq |

GPR179
Genomic Map