Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004220.hg.1 |
TC01000274.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004220.hg.1 |
ENST00000344642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004220.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01004220.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000484271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000484271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000484271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000484271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150460.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150460.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150460.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150460.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150460.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100150460.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100150460.hg.1 |
TC0100007254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000344642.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000484271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000484271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000484271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000484271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
ENST00000543870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007254.hg.1 |
NM_001013693.2 |
Refseq |