Assay Details
| Gene Symbol: UPK3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182684.2 | NP_872625.1 | 3 - 4 | 694 | 82 | |
| GenBank mRNA | AY233462.1 | - | 3 - 4 | 527 | 82 | |
| BU150542.1 | - | 689 | 82 | |||
| Gene Symbol: LOC105375355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006716140.3 | XP_006716203.1 | 3 - 4 | 772 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003310.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000447046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000447046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000447046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000469114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000469114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000469114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000469114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000490360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000490360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | ENST00000490360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006882.hg.1 | ENST00000334348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006882.hg.1 | ENST00000443097.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006882.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006882.hg.1 | TC07003310.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000257632.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000394849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000447046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000447046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000447046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000469114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000469114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000469114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000469114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000490360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000490360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | ENST00000490360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008096.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156203.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156203.hg.1 | ENST00000469114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156203.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156203.hg.1 | ENST00000334348.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156203.hg.1 | ENST00000469114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700156203.hg.1 | NM_182684.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700156203.hg.1 | TC0700008096.hg.1 | Clariom D TC |

UPK3B
Genomic Map