Assay Details
| Gene Symbol: GANC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198141.2 | NP_937784.2 | 10 - 11 | 1376 | 89 | |
| GenBank mRNA | AF525397.1 | - | 6 - 7 | 629 | 89 | |
| AF545045.1 | - | 10 - 11 | 1070 | 89 | ||
| AF545046.1 | - | 10 - 11 | 1070 | 89 | ||
| AK074037.1 | - | 10 - 11 | 1268 | 89 | ||
| BC059406.1 | - | 10 - 11 | 1248 | 30330796 | 89 | |
| BC093833.1 | - | 10 - 11 | 1093 | 7939678 | 89 | |
| BC112051.1 | - | 10 - 11 | 1093 | 8327519 | 89 | |
More Information
Set Membership: |
|

GANC
Genomic Map