| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15002535.hg.1 |
ENST00000318010.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15002535.hg.1 |
NM_198141.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15002535.hg.1 |
TC15002792.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15002536.hg.1 |
ENST00000318010.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15002536.hg.1 |
NM_198141.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15002536.hg.1 |
TC15002792.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1500181919.hg.1 |
NM_198141.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500181919.hg.1 |
TC1500010724.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500181919.hg.1 |
NM_198141.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500181920.hg.1 |
NM_198141.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500181920.hg.1 |
TC1500010724.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500181920.hg.1 |
NM_198141.2 |
Refseq |