Assay Details
| Gene Symbol: CYP3A43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012542.1 | XP_016868031.1 | 11 | 1492 | 132 | |
| XM_017012543.1 | XP_016868032.1 | 10 | 1650 | 132 | ||
| XM_017012544.1 | XP_016868033.1 | 9 | 1378 | 132 | ||
| XM_017012545.1 | XP_016868034.1 | 9 | 1358 | 132 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CYP3A43
Genomic Map