Assay Details
| Gene Symbol: ABCC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003087.1 | 6 | 1820 | 96 | ||
| GenBank mRNA | AY063514.1 | - | 6 | 1700 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RNA binding motif protein 11 | RBM11 | -71.1kb | Sense | Chr. 21 - 14216107 - 14228372 on NCBI Build GRCh38 | |
| Downstream | heat shock protein family A (Hsp70) member 13 | HSPA13 | STCH | 69.7kb | Anti-sense | Chr. 21 - 14371115 - 14383188 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ABCC13
Genomic Map