| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419349.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000419783.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000496791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
ENST00000620890.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_000581.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011059.hg.1 |
NM_201397.2 |
Refseq |