Assay Details
| Gene Symbol: OCLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205254.1 | NP_001192183.1 | 3 | 807 | 101 | |
| NM_002538.3 | NP_002529.1 | 3 | 1037 | 101 | ||
| XM_017008913.1 | XP_016864402.1 | 3 | 807 | 101 | ||
| XM_017008914.1 | XP_016864403.1 | 3 | 782 | 101 | ||
| GenBank mRNA | AB451306.1 | - | 2 | 601 | 101 | |
| AB451437.1 | - | 2 | 601 | 101 | ||
| AK290697.1 | - | 3 | 769 | 101 | ||
| AK313501.1 | - | 3 | 780 | 101 | ||
| BC029886.1 | - | 3 | 779 | 5179203 | 101 | |
| GQ402517.1 | - | 2 | 645 | 101 | ||
| U49184.1 | - | 3 | 768 | 101 | ||
| U53823.1 | - | 3 | 1038 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

OCLN
Genomic Map