Assay Details
| Gene Symbol: RTEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283009.1 | NP_001269938.1 | 3 | 1046 | 66 | |
| NM_001283010.1 | NP_001269939.1 | 2 | 767 | 66 | ||
| NM_016434.3 | NP_057518.1 | 3 | 1046 | 66 | ||
| NM_032957.4 | NP_116575.3 | 3 | 1046 | 66 | ||
| GenBank mRNA | AF217795.1 | - | 3 | 1046 | 66 | |
| AK299332.1 | - | 3 | 541 | 66 | ||
| AK302508.1 | - | 3 | 565 | 66 | ||
| AK304798.1 | - | 2 | 263 | 66 | ||
| JX119186.1 | - | 2 | 219 | 66 | ||
| Gene Symbol: RTEL1-TNFRSF6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037882.1 | 3 | 1046 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK304798.1, NM_001283010.1 transcripts.
Set Membership: |
|

RTEL1
Genomic Map