Assay Details
| Gene Symbol: EIF1AX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001412.3 | NP_001403.1 | 6 - 7 | 631 | 163 | |
| GenBank mRNA | BC000793.1 | - | 6 - 7 | 592 | 3451631 | 163 |
| BC067851.1 | - | 6 - 7 | 618 | 5288453 | 163 | |
| BC110421.1 | - | 6 - 7 | 596 | 163 | ||
| L18960.1 | - | 6 - 7 | 630 | 163 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000909.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012284.hg.1 | ENST00000379607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012284.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012284.hg.1 | TC0X000909.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012289.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012289.hg.1 | ENST00000379607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012289.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012289.hg.1 | TC0X000909.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012281.hg.1 | ENST00000379607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012281.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012281.hg.1 | TC0X000909.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012291.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012291.hg.1 | ENST00000379607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012291.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012291.hg.1 | TC0X000909.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012280.hg.1 | ENST00000379607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012280.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012280.hg.1 | TC0X000909.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012286.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012286.hg.1 | ENST00000379607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012286.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012286.hg.1 | TC0X000909.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012285.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012285.hg.1 | ENST00000379607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012285.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012285.hg.1 | TC0X000909.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012290.hg.1 | ENST00000379607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012290.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012290.hg.1 | TC0X000909.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018410.hg.1 | ENST00000419267.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012287.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012287.hg.1 | ENST00000379607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012287.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012287.hg.1 | TC0X000909.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012288.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012288.hg.1 | ENST00000379607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012288.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X012288.hg.1 | TC0X000909.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009215.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163342.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163342.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163342.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163342.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163342.hg.1 | TC0X00009215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00163335.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163335.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163335.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163335.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163335.hg.1 | TC0X00009215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00163341.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163341.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163341.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163341.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163341.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163341.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163341.hg.1 | TC0X00009215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00163334.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163334.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163334.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163334.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163334.hg.1 | TC0X00009215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00163336.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163336.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163336.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163336.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163336.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163336.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163336.hg.1 | TC0X00009215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018410.hg.1 | ENST00000419267.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018410.hg.1 | ENST00000624828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163339.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163339.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163339.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163339.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163339.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163339.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163339.hg.1 | TC0X00009215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00163340.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163340.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163340.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163340.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163340.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163340.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163340.hg.1 | TC0X00009215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00163344.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163344.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163344.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163344.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163344.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163344.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163344.hg.1 | TC0X00009215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00163333.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163333.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163333.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163333.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163333.hg.1 | TC0X00009215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00163338.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163338.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163338.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163338.hg.1 | ENST00000379593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163338.hg.1 | ENST00000379607.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00163338.hg.1 | NM_001412.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00163338.hg.1 | TC0X00009215.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 142269 | - | - |

EIF1AX
Genomic Map