Assay Details
| Gene Symbol: SPINK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195290.1 | NP_001182219.1 | 2 - 3 | 239 | 72 | |
| NM_205841.3 | NP_995313.2 | 1 - 2 | 366 | 72 | ||
| GenBank mRNA | AY358716.1 | - | 1 - 2 | 86 | 72 | |
| BC032003.1 | - | 1 - 2 | 319 | 4754193 | 72 | |
| BE122852.1 | - | 1 - 2 | 155 | 72 | ||
| GQ504704.1 | - | 1 - 2 | 384 | 72 | ||
| GQ504705.1 | - | 1 - 2 | 320 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000807.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011473.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011473.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011473.hg.1 | TC05000807.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011471.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011471.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011471.hg.1 | TC05000807.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011474.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011474.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011474.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011474.hg.1 | TC05000807.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011472.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011472.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011472.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011472.hg.1 | TC05000807.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500162896.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162896.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162896.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162896.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162896.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162896.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162896.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162896.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162896.hg.1 | TC0500009033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500162897.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162897.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162897.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162897.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162897.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162897.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162897.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162897.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162897.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162897.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162897.hg.1 | TC0500009033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009033.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162898.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162898.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162898.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162898.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162898.hg.1 | TC0500009033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500162900.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162900.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162900.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162900.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162900.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162900.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162900.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162900.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162900.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162900.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162900.hg.1 | TC0500009033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500162899.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162899.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162899.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162899.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162899.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162899.hg.1 | ENST00000325630.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162899.hg.1 | ENST00000514389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162899.hg.1 | ENST00000621437.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162899.hg.1 | NM_001195290.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162899.hg.1 | NM_205841.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162899.hg.1 | TC0500009033.hg.1 | Clariom D TC |

SPINK6
Genomic Map