| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003090.hg.1 |
ENST00000381440.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003090.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003090.hg.1 |
TC16000210.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003089.hg.1 |
ENST00000381440.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003089.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003089.hg.1 |
TC16000210.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003093.hg.1 |
ENST00000381440.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003093.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003093.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16003093.hg.1 |
TC16000210.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011359.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189591.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189591.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189591.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189591.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189591.hg.1 |
TC1600011359.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600189591.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189591.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189591.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189591.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189585.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189585.hg.1 |
TC1600011359.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600189585.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189589.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189589.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189589.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189589.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189589.hg.1 |
TC1600011359.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600189589.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189589.hg.1 |
ENST00000566735.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189589.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189589.hg.1 |
NR_028028.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189587.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189587.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189587.hg.1 |
TC1600011359.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600189587.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189587.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189586.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189586.hg.1 |
NM_001034841.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600189586.hg.1 |
TC1600011359.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600189586.hg.1 |
ENST00000381440.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600189586.hg.1 |
NM_001034841.3 |
Refseq |