Assay Details
| Gene Symbol: NUP62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193357.1 | NP_001180286.1 | 2 | 3001 | 90 | |
| NM_012346.4 | NP_036478.2 | 2 | 3042 | 90 | ||
| NM_016553.4 | NP_057637.2 | 3 | 3403 | 90 | ||
| NM_153718.3 | NP_714940.1 | 2 | 3081 | 90 | ||
| NM_153719.3 | NP_714941.1 | 3 | 3236 | 90 | ||
| GenBank mRNA | AF318357.1 | - | 3 | 1782 | 90 | |
| AL162061.1 | - | 2 | 3002 | 90 | ||
| BM672281.1 | - | 1 | 90 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NUP62
Genomic Map