Assay Details
| Gene Symbol: NUP62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193357.1 | NP_001180286.1 | 2 | 2985 | 90 | |
| NM_012346.4 | NP_036478.2 | 2 | 3026 | 90 | ||
| NM_016553.4 | NP_057637.2 | 3 | 3387 | 90 | ||
| NM_153718.3 | NP_714940.1 | 2 | 3065 | 90 | ||
| NM_153719.3 | NP_714941.1 | 3 | 3220 | 90 | ||
| GenBank mRNA | AF318357.1 | - | 3 | 1766 | 90 | |
| AL162061.1 | - | 2 | 2986 | 90 | ||
| BM672281.1 | - | 1 | 90 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19002608.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
RNA export from nucleus
mRNA export from nucleus
tRNA export from nucleus
protein import into nucleus
mitotic nuclear division
mitotic nuclear envelope disassembly
cell surface receptor signaling pathway
spermatogenesis
cell aging
cell death
negative regulation of cell proliferation
hormone-mediated signaling pathway
regulation of signal transduction
regulation of glucose transport
viral process
protein sumoylation
viral transcription
gene silencing by RNA
negative regulation of epidermal growth factor receptor signaling pathway
regulation of protein import into nucleus
negative regulation of apoptotic process
negative regulation of programmed cell death
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of MAP kinase activity
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
regulation of Ras protein signal transduction
negative regulation of Ras protein signal transduction
protein heterotrimerization
ribonucleoprotein complex export from nucleus
intracellular transport of virus
regulation of cellular response to heat
chromatin binding
nucleocytoplasmic transporter activity
protein binding
phospholipid binding
structural constituent of nuclear pore
kinesin binding
receptor signaling complex scaffold activity
Hsp70 protein binding
SH2 domain binding
ubiquitin binding
thyroid hormone receptor binding
PTB domain binding
Hsp90 protein binding
transcription, DNA-templated
RNA export from nucleus
mRNA export from nucleus
tRNA export from nucleus
protein import into nucleus
mitotic nuclear division
mitotic nuclear envelope disassembly
cell surface receptor signaling pathway
spermatogenesis
cell aging
cell death
negative regulation of cell proliferation
hormone-mediated signaling pathway
regulation of signal transduction
regulation of glucose transport
viral process
protein sumoylation
viral transcription
gene silencing by RNA
negative regulation of epidermal growth factor receptor signaling pathway
regulation of protein import into nucleus
negative regulation of apoptotic process
negative regulation of programmed cell death
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of MAP kinase activity
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
regulation of Ras protein signal transduction
negative regulation of Ras protein signal transduction
protein heterotrimerization
ribonucleoprotein complex export from nucleus
intracellular transport of virus
regulation of cellular response to heat
chromatin binding
nucleocytoplasmic transporter activity
protein binding
phospholipid binding
structural constituent of nuclear pore
kinesin binding
receptor signaling complex scaffold activity
Hsp70 protein binding
SH2 domain binding
ubiquitin binding
thyroid hormone receptor binding
PTB domain binding
Hsp90 protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19023635.hg.1 | ENST00000319225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023635.hg.1 | ENST00000352066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023635.hg.1 | ENST00000413454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023635.hg.1 | ENST00000422090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023635.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023635.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023635.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023635.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023635.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023635.hg.1 | TC19002608.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023636.hg.1 | ENST00000352066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023636.hg.1 | ENST00000413454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023636.hg.1 | ENST00000422090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023636.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023636.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023636.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023636.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023636.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023636.hg.1 | TC19002608.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023634.hg.1 | ENST00000319225.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023634.hg.1 | ENST00000352066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023634.hg.1 | ENST00000413454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023634.hg.1 | ENST00000422090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023634.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023634.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023634.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023634.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023634.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023634.hg.1 | TC19002608.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023638.hg.1 | ENST00000352066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023638.hg.1 | ENST00000413454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023638.hg.1 | ENST00000422090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023638.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023638.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023638.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023638.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023638.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023638.hg.1 | TC19002608.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023637.hg.1 | ENST00000352066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023637.hg.1 | ENST00000413454.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023637.hg.1 | ENST00000422090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023637.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023637.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023637.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023637.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023637.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023637.hg.1 | TC19002608.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | ENST00000413454.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | ENST00000422090.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | TC1900012007.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | ENST00000413454.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | ENST00000422090.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207299.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | ENST00000352066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | ENST00000413454.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | ENST00000422090.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | TC1900012007.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | ENST00000352066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | ENST00000413454.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | ENST00000422090.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207301.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | ENST00000352066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | ENST00000422090.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | TC1900012007.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | ENST00000352066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | ENST00000422090.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207303.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | ENST00000352066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | ENST00000422090.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | ENST00000596217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | ENST00000597029.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | ENST00000597723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | TC1900012007.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | ENST00000352066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | ENST00000422090.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | ENST00000596217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | ENST00000597029.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | ENST00000597723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207305.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | ENST00000352066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | ENST00000422090.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | NM_153719.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | TC1900012007.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | ENST00000352066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | ENST00000422090.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | NM_001193357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | NM_012346.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | NM_016553.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | NM_153718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900207302.hg.1 | NM_153719.3 | Refseq |

NUP62
Genomic Map