Assay Details
| Gene Symbol: NUP62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193357.1 | NP_001180286.1 | 2 | 3005 | 119 | |
| NM_012346.4 | NP_036478.2 | 2 | 3046 | 119 | ||
| NM_016553.4 | NP_057637.2 | 3 | 3407 | 119 | ||
| NM_153718.3 | NP_714940.1 | 2 | 3085 | 119 | ||
| NM_153719.3 | NP_714941.1 | 3 | 3240 | 119 | ||
| GenBank mRNA | AF318357.1 | - | 3 | 1786 | 119 | |
| AL162061.1 | - | 2 | 3006 | 119 | ||
| BM672281.1 | - | 1 | 119 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NUP62
Genomic Map