Assay Details
| Gene Symbol: EIF3J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284335.1 | NP_001271264.1 | 4 - 5 | 415 | 80 | |
| NM_003758.3 | NP_003749.2 | 4 - 5 | 415 | 80 | ||
| GenBank mRNA | AK002006.1 | - | 4 - 5 | 335 | 80 | |
| AK023388.1 | - | 4 - 5 | 365 | 80 | ||
| AK300659.1 | - | 4 - 5 | 300 | 80 | ||
| BC002719.2 | - | 4 - 5 | 348 | 3632801 | 80 | |
| U97670.1 | - | 4 - 5 | 348 | 80 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300009154.hg.1 | ENST00000466044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009154.hg.1 | ENST00000466044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009154.hg.1 | ENST00000466044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000536248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000536248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000536248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000536248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000261868.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000424492.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000535391.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000536248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000536248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000536248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000536248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | ENST00000558353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_001284336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007093.hg.1 | NM_003758.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009154.hg.1 | ENST00000466044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009154.hg.1 | ENST00000466044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009154.hg.1 | ENST00000466044.1 | Ensembl |

EIF3J
Genomic Map