Assay Details
| Gene Symbol: NTRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007097.2 | NP_001007098.1 | 13 | 3207 | 108 | |
| NM_001291937.1 | NP_001278866.1 | 15 | 3618 | 108 | ||
| XM_005252007.3 | XP_005252064.1 | 14 | 3538 | 108 | ||
| XM_017014758.1 | XP_016870247.1 | 15 | 3593 | 108 | ||
| XM_017014759.1 | XP_016870248.1 | 14 | 3354 | 108 | ||
| XM_017014760.1 | XP_016870249.1 | 15 | 4080 | 108 | ||
| XM_017014761.1 | XP_016870250.1 | 13 | 3254 | 108 | ||
| GenBank mRNA | AF508964.1 | - | 12 | 2724 | 108 | |
| AL713745.1 | - | 1 | 441 | 108 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NTRK2
Genomic Map