Assay Details
| Gene Symbol: RAB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017817.2 | NP_060287.1 | 1 - 2 | 392 | 80 | |
| GenBank mRNA | AJ272065.1 | - | 1 - 2 | 174 | 80 | |
| AK000436.1 | - | 1 - 2 | 379 | 80 | ||
| AK000446.1 | - | 1 - 2 | 80 | |||
| BC026025.1 | - | 1 - 2 | 388 | 4636491 | 80 | |
| BU528103.1 | - | 392 | 80 | |||
| HY010547.1 | - | 1 - 2 | 347 | 80 | ||
More Information
Set Membership: |
|

RAB20
Genomic Map