Assay Details
| Gene Symbol: GUSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000181.3 | NP_000172.2 | 11 - 12 | 1919 | 149 | |
| NM_001284290.1 | NP_001271219.1 | 9 - 10 | 1481 | 149 | ||
| NM_001293104.1 | NP_001280033.1 | 10 - 11 | 1734 | 149 | ||
| NM_001293105.1 | NP_001280034.1 | 9 - 10 | 1591 | 149 | ||
| NR_120531.1 | 10 - 11 | 1834 | 149 | |||
| XM_005250297.3 | XP_005250354.1 | 10 - 11 | 1761 | 149 | ||
| XM_011516113.1 | XP_011514415.1 | 10 - 11 | 1663 | 149 | ||
| XM_011516114.1 | XP_011514416.1 | 10 - 11 | 1591 | 149 | ||
| XM_017012091.1 | XP_016867580.1 | 9 - 10 | 1510 | 149 | ||
| XM_017012092.1 | XP_016867581.1 | 9 - 10 | 1497 | 149 | ||
| XM_017012093.1 | XP_016867582.1 | 9 - 10 | 1438 | 149 | ||
| GenBank mRNA | AK096764.1 | - | 3 - 4 | 2177 | 149 | |
| AK223406.1 | - | 11 - 12 | 1818 | 149 | ||
| AK300223.1 | - | 11 - 12 | 1708 | 149 | ||
| AK303819.1 | - | 9 - 10 | 1378 | 149 | ||
| AK312752.1 | - | 11 - 12 | 1835 | 149 | ||
| BC014142.2 | - | 11 - 12 | 1789 | 4662011 | 149 | |
| BM719712.1 | - | 2 - 3 | 172 | 149 | ||
| CD014092.1 | - | 10 - 11 | 1616 | 149 | ||
| CD014093.1 | - | 9 - 10 | 1474 | 149 | ||
| CD014094.1 | - | 10 - 11 | 1716 | 149 | ||
| M15182.1 | - | 11 - 12 | 1814 | 149 | ||
More Information
Set Membership: |
|

GUSB
Genomic Map