Assay Details
| Gene Symbol: CXCL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000609.6 | NP_000600.1 | 1 - 2 | 157 | 100 | |
| NM_001033886.2 | NP_001029058.1 | 1 - 2 | 157 | 100 | ||
| NM_001178134.1 | NP_001171605.1 | 1 - 2 | 157 | 100 | ||
| NM_001277990.1 | NP_001264919.1 | 1 - 2 | 157 | 100 | ||
| NM_199168.3 | NP_954637.1 | 1 - 2 | 157 | 100 | ||
| GenBank mRNA | AK292628.1 | - | 1 - 2 | 160 | 100 | |
| AK311814.1 | - | 1 - 2 | 155 | 100 | ||
| AU120056.1 | - | 1 - 2 | 152 | 100 | ||
| AY644456.1 | - | 1 - 2 | 114 | 100 | ||
| AY874118.1 | - | 1 - 2 | 144 | 100 | ||
| BC031072.1 | - | 1 - 2 | 149 | 100 | ||
| BC039893.1 | - | 1 - 2 | 135 | 5729604 | 100 | |
| CD172464.1 | - | 1 - 2 | 155 | 100 | ||
| DA266752.1 | - | 1 - 2 | 158 | 100 | ||
| DQ345517.1 | - | 1 - 2 | 109 | 100 | ||
| DQ345518.1 | - | 1 - 2 | 104 | 100 | ||
| DQ345519.1 | - | 1 - 2 | 109 | 100 | ||
| DQ345520.1 | - | 1 - 2 | 109 | 100 | ||
| L36033.1 | - | 1 - 2 | 144 | 100 | ||
| L36034.1 | - | 1 - 2 | 144 | 100 | ||
| U16752.1 | - | 1 - 2 | 145 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK292628.1, AK311814.1, AY644456.1, AY874118.1, BC039893.1, DQ345517.1, DQ345518.1, DQ345519.1, DQ345520.1, L36033.1, L36034.1, NM_000609.6, NM_001033886.2, NM_001178134.1, NM_001277990.1, NM_199168.3, U16752.1 transcripts.
Set Membership: |
|

CXCL12
Genomic Map