Assay Details
| Gene Symbol: MRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002438.3 | NP_002429.1 | 5 - 6 | 1040 | 102 | |
| GenBank mRNA | AK308132.1 | - | 5 - 6 | 1041 | 102 | |
| AK308167.1 | - | 5 - 6 | 1036 | 102 | ||
| BC142642.1 | - | 5 - 6 | 1021 | 102 | ||
| BC146838.1 | - | 5 - 6 | 963 | 9056765 | 102 | |
| DQ663787.1 | - | 5 - 6 | 917 | 102 | ||
| J05550.1 | - | 5 - 6 | 1020 | 102 | ||
| X55635.1 | - | 5 - 6 | 993 | 102 | ||
More Information
Set Membership: |
|

MRC1
Genomic Map