| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019252.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019252.hg.1 |
NM_004364.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019252.hg.1 |
TC19001396.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019253.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019253.hg.1 |
NM_004364.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19019253.hg.1 |
TC19001396.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_001285829.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_001285829.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_001285829.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_001287424.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_001287424.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_001287424.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_001287435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_001287435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_001287435.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_004364.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_004364.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010386.hg.1 |
NM_004364.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007772.hg.1 |
ENST00000587312.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007772.hg.1 |
ENST00000587312.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007772.hg.1 |
ENST00000587312.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007772.hg.1 |
ENST00000587312.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_001285829.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_001285829.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_001285829.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_001287424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_001287424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_001287424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_001287435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_001287435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_001287435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_004364.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_004364.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010386.hg.1 |
NM_004364.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185100.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185100.hg.1 |
NM_001285829.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185100.hg.1 |
NM_001287424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185100.hg.1 |
NM_001287435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185100.hg.1 |
NM_004364.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185100.hg.1 |
TC1900010386.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900185100.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185100.hg.1 |
NM_001285829.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185100.hg.1 |
NM_001287424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185100.hg.1 |
NM_001287435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185100.hg.1 |
NM_004364.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185102.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185102.hg.1 |
NM_001285829.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185102.hg.1 |
NM_001287424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185102.hg.1 |
NM_001287435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185102.hg.1 |
NM_004364.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185102.hg.1 |
TC1900010386.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900185102.hg.1 |
ENST00000498907.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900185102.hg.1 |
NM_001285829.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185102.hg.1 |
NM_001287424.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185102.hg.1 |
NM_001287435.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900185102.hg.1 |
NM_004364.4 |
Refseq |