Assay Details
| Gene Symbol: SRD5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000348.3 | NP_000339.2 | 2 - 3 | 511 | 99 | |
| XM_011533069.2 | XP_011531371.1 | 2 - 3 | 1226 | 99 | ||
| XM_011533072.2 | XP_011531374.1 | 5 - 6 | 656 | 99 | ||
| GenBank mRNA | AK307235.1 | - | 2 - 3 | 247 | 99 | |
| AK316597.1 | - | 2 - 3 | 481 | 99 | ||
| BC112252.1 | - | 2 - 3 | 498 | 8327720 | 99 | |
| BC113641.1 | - | 2 - 3 | 486 | 8322693 | 99 | |
| EF560740.1 | - | 2 - 3 | 479 | 99 | ||
| M74047.1 | - | 2 - 3 | 467 | 99 | ||
More Information
Set Membership: |
|

SRD5A2
Genomic Map