| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022034.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022034.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022034.hg.1 |
TC19001614.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000589594.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000589776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
ENST00000589776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022031.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022031.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022031.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022031.hg.1 |
TC19001614.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022033.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022033.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022033.hg.1 |
TC19001614.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022032.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022032.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022032.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022032.hg.1 |
TC19001614.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022028.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022028.hg.1 |
ENST00000429338.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022028.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022028.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022028.hg.1 |
TC19001614.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000589594.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000589776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
ENST00000589776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010925.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190644.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190644.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190644.hg.1 |
ENST00000589776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190644.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190644.hg.1 |
TC1900010925.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900190644.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190644.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190644.hg.1 |
ENST00000589776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190644.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190646.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190646.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190646.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190646.hg.1 |
TC1900010925.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900190646.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190646.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190646.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190648.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190648.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190648.hg.1 |
TC1900010925.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900190648.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190648.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190649.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190649.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190649.hg.1 |
TC1900010925.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900190649.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190649.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190647.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190647.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190647.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190647.hg.1 |
TC1900010925.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900190647.hg.1 |
ENST00000317951.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190647.hg.1 |
ENST00000438936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190647.hg.1 |
NM_198478.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190645.hg.1 |
ENST00000589776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190645.hg.1 |
TC1900010925.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900190645.hg.1 |
ENST00000589776.1 |
Ensembl |