Assay Details
| Gene Symbol: MT1IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003669.2 | 2 - 3 | 145 | 148 | ||
| GenBank mRNA | BC144614.1 | - | 2 - 3 | 129 | 148 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000475.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 967963 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3367293 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4234 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4235 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 40707 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3367293 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3367293 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035534 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035546 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575604 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16899591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035544 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035540 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035545 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16899574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035542 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16899571 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035533 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035549 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575604 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035547 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035534 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035549 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035545 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16899591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16899574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035542 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035546 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035540 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035544 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16899571 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30575608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10035533 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16899561 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035542 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035582 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035575 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035559 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035524 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035574 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035534 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035591 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035532 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035529 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035579 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035536 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035558 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035533 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035512 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035572 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16899554 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035569 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16899599 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035521 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035508 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035567 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035561 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035580 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035563 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035570 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16899601 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035526 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16899559 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035546 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035519 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035515 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035589 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035522 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035538 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035547 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035516 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16899591 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035571 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035543 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035527 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10035587 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_003669_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7995836 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7995835 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7995837 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7995834 | - | - |
| Affymetrix Human Genome U219 Array | 11763941_at | - | - |
| Affymetrix Human Genome U219 Array | 11762352_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11757246_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11756070_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11756070_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11763941_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11762352_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11757246_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC16000475.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC16000475.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC16000475.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC16000475.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC16000475.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC16000475.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | NR_104046.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | NR_104046.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007967.hg.1 | NR_104046.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006534.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006534.hg.1 | TC16000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006532.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006532.hg.1 | TC16000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006535.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006535.hg.1 | TC16000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006531.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006531.hg.1 | TC16000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006530.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006530.hg.1 | TC16000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006536.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16006536.hg.1 | TC16000475.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600158166.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158166.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158166.hg.1 | NR_104046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158166.hg.1 | TC1600007967.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600158166.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158166.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158166.hg.1 | NR_104046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158160.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600158160.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158160.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158160.hg.1 | TC1600007967.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600158160.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600158160.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158160.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158163.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600158163.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158163.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158163.hg.1 | NR_104046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158163.hg.1 | TC1600007967.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600158163.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600158163.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158163.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158163.hg.1 | NR_104046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158155.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600158155.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158155.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158155.hg.1 | NR_104046.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158155.hg.1 | TC1600007967.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600158155.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600158155.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158155.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600158155.hg.1 | NR_104046.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | ENST00000379816.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | NR_003669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | NR_104045.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | NR_104046.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | NR_104046.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007967.hg.1 | NR_104046.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054628 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054628 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3245209 | NR_003669.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3245209 | NR_003669.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2136089 | AF348997.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3245209 | NR_003669.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3245209 | NR_003669.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2136089 | AF348997.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2136089 | AF348997.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2136089 | AF348997.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2136089 | AF348997.1 | human_transcriptome |

MT1IP
Genomic Map