Assay Details
| Gene Symbol: MLLT10-PICALM fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF060930.1 | - | 97 | 119 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLLT10-PICALM fusion| Gene Symbol: MLLT10-PICALM fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF060930.1 | - | 97 | 119 | ||
Set Membership: |
|