Assay Details
| Gene Symbol: OAZ1-ULK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098316.1 | - | 188 | 152 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OAZ1-ULK2 fusion| Gene Symbol: OAZ1-ULK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098316.1 | - | 188 | 152 | ||
Set Membership: |
|