Assay Details
| Gene Symbol: LOC400464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127420.1 | - | 1 | 657 | 125 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09871037 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16832444 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16832452 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30375353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09871040 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16832473 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09871043 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30375361 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16832454 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09871045 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16832484 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16832457 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30375338 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09871044 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16832371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09871035 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16832456 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30375364 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 25216 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 680680 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 400371 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 401210 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 6137 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 154828 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 326827 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 36989 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 38399 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3342345 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P548953 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P548953 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 39631 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03097062 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03097062 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7986429 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7986430 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7986432 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7986431 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7986428 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010797.hg.1 | ENST00000560059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010797.hg.1 | ENST00000560059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010797.hg.1 | ENST00000560059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010797.hg.1 | ENST00000560059.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010797.hg.1 | ENST00000622345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010797.hg.1 | ENST00000622345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010797.hg.1 | ENST00000622345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010673.hg.1 | ENST00000558592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010673.hg.1 | ENST00000558592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010673.hg.1 | ENST00000558592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010673.hg.1 | ENST00000560292.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010673.hg.1 | ENST00000560292.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010673.hg.1 | ENST00000560292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010797.hg.1 | ENST00000560059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010797.hg.1 | ENST00000560059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010797.hg.1 | ENST00000560059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010797.hg.1 | ENST00000560059.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010797.hg.1 | ENST00000622345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010797.hg.1 | ENST00000622345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010797.hg.1 | ENST00000622345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010673.hg.1 | ENST00000558592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010673.hg.1 | ENST00000558592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010673.hg.1 | ENST00000558592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010673.hg.1 | ENST00000560292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010673.hg.1 | ENST00000560292.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010673.hg.1 | ENST00000560292.2 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 243957_at | - | - |
| Human Genome U133 Plus 2.0 Array | 243957_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243957_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243957_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243957_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243957_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243957_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243957_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243957_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243957_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243957_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243957_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 243957_at9 | - | - |
| Human Genome U95 Set | 49951_PM_at | - | - |
| Human Genome U95 Set | 49951_at | - | - |
| Human Genome U133 Set | 243957_PM_at | - | - |
| Human Genome U133 Set | 243957_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3307611 | AK127420.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2320480 | AK127420.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1786168 | AK127420.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1786168 | AK127420.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1676395 | - | - |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1786168 | AK127420.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2320480 | AK127420.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1676395 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1786168 | AK127420.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1676395 | - | - |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1786168 | AK127420.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1786168 | AK127420.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2320480 | AK127420.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1786168 | AK127420.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2320480 | AK127420.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2320480 | AK127420.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1786168 | AK127420.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1786168 | AK127420.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2320480 | AK127420.1 | human_transcriptome |

LOC400464
Genomic Map