Assay Details
| Gene Symbol: LOC650293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040071.1 | NP_001035160.1 | 1 | 567 | 134 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23070816 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 800787 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 74586 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3405535 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P352358 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 44395 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20950 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 51500 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3405535 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P352358 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3405535 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P352358 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P352358 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P427731 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P456046 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15543956 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02919516 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8149149 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149209 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7960899 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8144423 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7960897 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7949897 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8144497 | - | - |
| Affymetrix Human Genome U219 Array | 11715160_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11715160_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 215770_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216476_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216476_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 215770_at | - | - |
| Human Genome U133A 2.0 Array | 215770_PM_at | - | - |
| Human Genome U133A 2.0 Array | 215770_at | - | - |
| Human Genome U133A 2.0 Array | 216476_PM_at | - | - |
| Human Genome U133A 2.0 Array | 216476_at | - | - |
| Human Genome U133 Set | 216476_PM_at | - | - |
| Human Genome U133 Set | 216476_at | - | - |
| Human Genome U133 Set | 215770_at | - | - |
| Human Genome U133 Set | 215770_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025257 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025257 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1685813 | - | - |
| HumanHT-12 v4 Expression BeadChip | ILMN_2079225 | NM_001040071.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2079225 | AW009208.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1685813 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_2079225 | NM_001040071.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2079225 | AW009208.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2079225 | NM_001040071.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2079225 | AW009208.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2079225 | NM_001040071.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2079225 | AW009208.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1685813 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2079225 | NM_001040071.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2079225 | AW009208.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2079225 | NM_001040071.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2079225 | AW009208.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1685813 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1685813 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2079225 | NM_001040071.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2079225 | AW009208.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2079225 | NM_001040071.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2079225 | AW009208.1 | human_transcriptome |

LOC650293
Genomic Map