Assay Details
| Gene Symbol: NBR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003108.1 | 3 - 4 | 522 | 154 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000547.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000464237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000464237.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | ENST00000587322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006668.hg.1 | ENST00000356906.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006668.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006668.hg.1 | TC17000547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006669.hg.1 | ENST00000356906.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006669.hg.1 | ENST00000464237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006669.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006669.hg.1 | TC17000547.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000464237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000464237.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000467245.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | ENST00000587322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007939.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700158018.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700158018.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700158018.hg.1 | NR_003108.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700158018.hg.1 | TC1700007939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700158018.hg.1 | ENST00000356906.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700158018.hg.1 | ENST00000460115.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700158018.hg.1 | NR_003108.2 | Refseq |

NBR2
Genomic Map