| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008842.hg.1 |
ENST00000426688.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012187.hg.1 |
ENST00000427498.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010117.hg.1 |
ENST00000498116.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011381.hg.1 |
ENST00000486625.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600011119.hg.1 |
ENST00000484096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008795.hg.1 |
ENST00000475257.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010094.hg.1 |
ENST00000462503.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005540.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005540.hg.1 |
TC06000516.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005542.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005542.hg.1 |
ENST00000467020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005542.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005542.hg.1 |
TC06000516.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007525.hg.1 |
ENST00000479243.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009760.hg.1 |
ENST00000465163.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010310.hg.1 |
ENST00000490023.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011346.hg.1 |
ENST00000472730.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005541.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005541.hg.1 |
ENST00000467020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005541.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06005541.hg.1 |
TC06000516.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0800010094.hg.1 |
ENST00000462503.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010117.hg.1 |
ENST00000498116.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008795.hg.1 |
ENST00000475257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007525.hg.1 |
ENST00000479243.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600011119.hg.1 |
ENST00000484096.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012187.hg.1 |
ENST00000427498.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010310.hg.1 |
ENST00000490023.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154076.hg.1 |
TC0600007800.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100008842.hg.1 |
ENST00000426688.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011346.hg.1 |
ENST00000472730.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009760.hg.1 |
ENST00000465163.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011381.hg.1 |
ENST00000486625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154078.hg.1 |
TC0600007800.hg.1 |
Clariom D TC |