| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007800.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
ENST00000322203.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
ENST00000464112.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
ENST00000467020.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
ENST00000478340.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
ENST00000490335.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
NM_007104.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600154082.hg.1 |
TC0600007800.hg.1 |
Clariom D TC |