Assay Details
| Gene Symbol: EIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005801.3 | NP_005792.1 | 3 - 4 | 459 | 126 | |
| GenBank mRNA | AF083441.1 | - | 3 - 4 | 442 | 126 | |
| AF100737.1 | - | 3 - 4 | 427 | 126 | ||
| AJ012375.1 | - | 3 - 4 | 426 | 126 | ||
| AK311209.1 | - | 2 - 3 | 280 | 126 | ||
| AL050005.1 | - | 1 - 2 | 1306 | 126 | ||
| BC005118.1 | - | 3 - 4 | 439 | 2823520 | 126 | |
| BC008710.1 | - | 3 - 4 | 433 | 3350981 | 126 | |
| BU165055.1 | - | 3 - 4 | 476 | 126 | ||
| L26247.1 | - | 3 - 4 | 434 | 126 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000514.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000586699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000586699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000586699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006130.hg.1 | ENST00000310837.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006130.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006130.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006130.hg.1 | TC17000514.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | TC1700007851.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157010.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000310837.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000462917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000469308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000482111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000586699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000586699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000586699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | ENST00000591776.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | NM_005801.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007851.hg.1 | NM_005801.3 | Refseq |

EIF1
Genomic Map