Assay Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138771.3 | NP_620126.2 | 3 - 4 | 695 | 89 | |
| XM_017012775.1 | XP_016868264.1 | 4 - 5 | 935 | 89 | ||
| XM_017012776.1 | XP_016868265.1 | 3 - 4 | 678 | 89 | ||
| GenBank mRNA | AK026684.1 | - | 1 - 2 | 192 | 89 | |
| AK289911.1 | - | 3 - 4 | 671 | 89 | ||
| AY358713.1 | - | 3 - 4 | 662 | 89 | ||
| BC012427.1 | - | 1 - 2 | 276 | 3858521 | 89 | |
| BC104638.1 | - | 3 - 4 | 587 | 6620343 | 89 | |
More Information
Set Membership: |
|

CCDC126
Genomic Map